Flowline DataView Advanced Meter & Controller
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
23002-18 23002-20 23002-22
MFLX23002-18EA 1420 EUR
MFLX23002-18 MFLX23002-20 MFLX23002-22
Flowline DataView Advanced Meter & Controller
Niveausensoren
Easy configuration via push button display module.
  • Bright 6-digit LED main display and secondary unit display with relay status indicators
  • 1/8 DIN enclosure with NEMA 4X faceplate and shallow depth case
  • 32-point linearization function for volumetric tank measurement
  • Non-volatile memory with security password protected lock out
  • Isolated 4-20 mA repeater output with 40 mA internal power supply
The general purpose meter displays engineering units with one 4-20 mA or 0-10 VDC transmitter input channel and is offered in three configurations with optional 2More Product Information