Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX30544-20EA 202 EUR
MFLX30544-20
Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Afdichtingen Pakkingen
Easily sample fluid in critical high-purity system.
  • Three internal ports accept thermocouple probe0
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.