CPC® ChemQuik® Quick-Disconnect Fittings, Panel-Mount Coupling Bodies
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX31603-05EA 296 EUR
MFLX31603-05
CPC® ChemQuik® Quick-Disconnect Fittings, Panel-Mount Coupling Bodies
Fittingen en connectoren
All plastic construction for high-purity use.
  • 100% metal-free couplings are ideal for ultrapure fluids, deionized water, and aggressive chemical applications
These ChemQuik® quick-disconnect couplings provide easy single-handed disconnection. These couplings feature valves to provide flow shut-off. The all plastic construction and lubricant-free design enables use in the most demanding applications.
For a complete coupling, order one body and one insert of the same size.