CPC® Sanitary Steam-in-Place Connectors, Single Cycle
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31604-18 31604-20 31604-10 31604-18BLK 31604-20BLK 31604-10BLK 31309-61BLK
MFLX31604-18EA 305 EUR
MFLX31604-18 MFLX31604-20 MFLX31604-10 MFLX31604-18BLK MFLX31604-20BLK MFLX31604-10BLK MFLX31309-61BLK
CPC® Sanitary Steam-in-Place Connectors, Single Cycle
Fittingen en connectoren
Ensure a sterile connection.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • One disposable connector for steam sterilization and microbial process
  • Reduces down time and cleaning expenses
As part of a single-use system, these CPC® sanitary Steam-in-Place disposable connectors enhance the sterility of your bioprocessing line. The innovative product design allows for a one-step process for sterilization and media transfer while eliminating dead spots, reducing the number of connections, and eliminating the need for a laminar flow hood.More Product Information