McMillan Flow High-Accuracy Flowmeter Power Supply Adapters
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32704-55 32704-56
MFLX32704-55EA 153 EUR
MFLX32704-55 MFLX32704-56
McMillan Flow High-Accuracy Flowmeter Power Supply Adapters
Stroombronnen
Ideal for use with McMillan Flow High-Accuracy Flowmeters.
  • A wide range of output options for integration into any system
These power supplies are required to power the Modular Liquid Flow Rate Sensor Systems.