Masterflex® Barbed Low Flow Female Quick-Disconnect Fittings, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX40250-00EA 94 EUR
MFLX40250-00 MFLX40250-01 MFLX40250-08 MFLX40250-09 MFLX40250-15 MFLX40250-16 MFLX40250-23 MFLX40250-24 MFLX40250-30 MFLX40250-31 MFLX40250-38 MFLX40250-39 MFLX40250-45 MFLX40250-46 MFLX40250-53 MFLX40250-54
Masterflex® Barbed Low Flow Female Quick-Disconnect Fittings, Avantor®
Fittingen en connectoren
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Low Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured, aMore Product Information