Masterflex® Large Bore Male Luer to Barb Fitting, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX40313-01EA 20.7 EUR
MFLX40313-01 MFLX40313-02 MFLX40313-03 MFLX40313-04 MFLX40313-05 MFLX40313-06 MFLX40313-08 MFLX40313-09 MFLX40313-10 MFLX40313-11 MFLX40313-12 MFLX40313-13 MFLX40313-15 MFLX40313-16 MFLX40313-18 MFLX40313-20 MFLX40313-21
Masterflex® Large Bore Male Luer to Barb Fitting, Avantor®
Fittingen en connectoren
These large-bore adapters are perfect for handling higher flow volumes in process while still using a slip-lock-style fitting system.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing anMore Product Information