Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73007-02 73007-00 73007-11
MFLX73007-02EA 5760 EUR
MFLX73007-02 MFLX73007-00 MFLX73007-11
Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Pompen Pump Heads Gear Pump Heads
Smooth pulseless flow provides precise, leak-free delivery of fluids.
  • Compact size and easy to service
  • Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.