Eldex Low-Flow Piston Pump Systems
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
73150-02 73150-10 73150-12 73150-20 73150-40 73150-42 73150-50 73150-80
MFLX73150-02EA 6560 EUR
MFLX73150-02 MFLX73150-10 MFLX73150-12 MFLX73150-20 MFLX73150-40 MFLX73150-42 MFLX73150-50 MFLX73150-80
Eldex Low-Flow Piston Pump Systems
Pompen HPLC-pompen
Remote flow rate control via analog signal or RS232 interface.
  • Optional liquid ends for expanded flow range
  • Smooth fluid delivery from intra RPM stepper motor control
  • Integrated piston wash
Extended flow ranges are possible by using up to three interchangeable liquid end assemblies. Software control of refill rates and compressibility compensation add to the versatility of the pumps and enhance pump performance. Precise metering is attained by a positive-displacement reciprocating piston assembly. The high-resolution stepper motor is used toMore Product Information