Masterflex® I/P® MasterSense® Process Pumps with I/P® Rapid-Load® Pump Head
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MFLX77966-20EA 16050 EUR
MFLX77966-20 MFLX77966-22 MFLX77966-30 MFLX77966-32 MFLX77966-20-EU MFLX77966-22-EU MFLX77966-30-EU MFLX77966-32-EU
Masterflex® I/P® MasterSense® Process Pumps with I/P® Rapid-Load® Pump Head
Pompen Peristaltische pompen
Versatile pumps for process applications - with an intuitive touch-screen interface and advanced connectivity. This drive offers a broad flow range, from 0,0006 to 19 LPM, adaptable to a variety of applications and workflows. The drive housing is constructed of 316 stainless steel - IP 66 and NEMA 4X rated for washdown.
  • Powerful, space-efficient drives will accept all I/P® pump head and tubing options
  • I/P® Rapid-Load® pump head offers the best combination of pressure and flow performance
  • 17,8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and
More Product Information