Masterflex® Transfer Tubing, Tygon® A-60-F, Avantor®
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
95710-00 95710-10 95710-20 95710-40 95710-70 95710-90 95710-30 95710-50 95710-60 95710-80
MFLX95710-00EA 105 EUR
MFLX95710-00 MFLX95710-10 MFLX95710-20 MFLX95710-40 MFLX95710-70 MFLX95710-90 MFLX95710-30 MFLX95710-50 MFLX95710-60 MFLX95710-80
Masterflex® Transfer Tubing, Tygon® A-60-F, Avantor®
Slangen Transfer Tubing
Stay compliant with food applications.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for the food, dairy, and beverage industries. Characteristics include heat, ozone, and UV light resistant; non-aging; nonoxidizing; superior acid and alkali resistance. Tubing is manufactured accordingMore Product Information