Burkert Type 3233 316L Stainless Steel Diaphragm Valve
Leverancier: Avantor Fluid Handling
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
98516-28 98516-32 98516-58 98516-60 98516-62 98516-64 98516-66
MFLX98516-28EA 1000 EUR
MFLX98516-28 MFLX98516-32 MFLX98516-58 MFLX98516-60 MFLX98516-62 MFLX98516-64 MFLX98516-66
Burkert Type 3233 316L Stainless Steel Diaphragm Valve
Ventielen Diaphragm Valves
Fluids are hermetically sealed from operating system.
  • Zero dead volume
  • Autoclavable
Choose from EPDM or PTFE wetted materials and sanitary or weld end connections. These compact, manually-operated diaphragm valves consist of a flow-optimized stainless steel body, diaphragm, and a manual actuator. The diaphragm is both a switch element and a sealing element and can be easily replaced. The valves have no dead volume and can be mounted to be self-draining — this enables high flow capacities and a variety of applications to be realized. The flow can beMore Product Information