String mops, NovaKnit™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VW51-31-12W MD5131-12WIR MP5231-12WIR MP52-31-12W
MICRVW51-31-12WEA 34.3 EUR
MICRVW51-31-12W MICRMD5131-12WIR MICRMP5231-12WIR MICRMP52-31-12W
String mops, NovaKnit™
Moppen
A series of robust and absorbent tubular knit string mops for cleaning and liquid pick-up in controlled and critical environments. Available with a choice of durable, reusable fabrics.
  • Robust, tubular knit floor mop
  • Ribbed strands increase absorbency
  • Looped design with no exposed edges to ensure low particulates
  • More absorbent and durable than traditional cleanroom mops
  • Use with MH-1 handle or QDMH-1 MicroMop adapter with SSU series handles
  • Available gamma irradiated for aseptic environments
  • Holds up to strong disinfectants
  • Launderable for reuse
More Product Information