HPLC columns, Cogent HPS™ C8
Leverancier: MICROSOLV
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
75008-05P-2 75008-10P 75008-10P-2 75008-15P 75008-15P-2 75008-25P 75008-P100 75008-P150 75008-P250 75008-P50 75008-SP100 75008-SP150 75008-SP250 75008-SP50
MICS75008-05P-2EA 868 EUR
MICS75008-05P-2 MICS75008-10P MICS75008-10P-2 MICS75008-15P MICS75008-15P-2 MICS75008-25P MICS75008-P100 MICS75008-P150 MICS75008-P250 MICS75008-P50 MICS75008-SP100 MICS75008-SP150 MICS75008-SP250 MICS75008-SP50
HPLC columns, Cogent HPS™ C8
Kolommen voor chromatografie
Cogent HPS™ C8 columns are high-purity type-B, silica-based HPLC columns and have narrow particle distribution making them extremely high-quality.
  • Double endcapped
  • Features monofunctional bonding which provides greater stability and reproducibility
These type-B silica columns are made with extremely low metal content and are excellent choices for all types of analytes including bases, neutrals and acids by reversed phase or normal phase.