Glass Fibre Filters without Binder
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
APFA03700 APFF03700
MILFAPFA03700EA 59.3 EUR
MILFAPFA03700 MILFAPFF03700
Glass Fibre Filters without Binder
Filters Glasvezelfilters
Millipore fibrous disc filters are available in a wide range of flow rates and throughput capacities. Our glass fibre filters are available with or without binder resins and can be sterilised by EO, gamma, or autoclave (121 °C at 1 bar).
Glass fibre filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
Type APFA:
Retains fine particles with good efficiency, even at high flow rates
Recommended for monitoring wastewater and collecting suspendedMore Product Information