Ultracel PL Ultrafiltration Membrane Disc Filters
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PLAC02510 PLAC04310 PLAC04710 PLAC06210 PLAC07610 PLAC09005 PLAC15005 PLBC02510 PLBC04310 PLBC04710 PLBC06210 PLBC07610 PLBC09005 PLBC15005 PLCC02510 PLCC04310 PLCC04710 PLCC06210 PLCC07610 PLCC09005 PLCC15005 PLGC02510 PLGC04310 PLGC04710 PLGC06210 PLGC07610 PLGC09005 PLHK02510 PLHK04310 PLHK04710 PLHK06210 PLHK07610 PLHK09005 PLHK15005 PLTK02510 PLTK04310 PLTK04710 PLTK06210 PLTK09005 PLTK15005 PLTK07610 PLGC15005
MILFPLAC02510EA 214 EUR
MILFPLAC02510 MILFPLAC04310 MILFPLAC04710 MILFPLAC06210 MILFPLAC07610 MILFPLAC09005 MILFPLAC15005 MILFPLBC02510 MILFPLBC04310 MILFPLBC04710 MILFPLBC06210 MILFPLBC07610 MILFPLBC09005 MILFPLBC15005 MILFPLCC02510 MILFPLCC04310 MILFPLCC04710 MILFPLCC06210 MILFPLCC07610 MILFPLCC09005 MILFPLCC15005 MILFPLGC02510 MILFPLGC04310 MILFPLGC04710 MILFPLGC06210 MILFPLGC07610 MILFPLGC09005 MILFPLHK02510 MILFPLHK04310 MILFPLHK04710 MILFPLHK06210 MILFPLHK07610 MILFPLHK09005 MILFPLHK15005 MILFPLTK02510 MILFPLTK04310 MILFPLTK04710 MILFPLTK06210 MILFPLTK09005 MILFPLTK15005 MILFPLTK07610 MILFPLGC15005
Ultracel PL Ultrafiltration Membrane Disc Filters
Filters Membraanfilters
Ultrafiltration cut disc membranes are available in Ultracel® regenerated cellulose and Biomax® polyethersulvone (PES).
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic and tight microstructure assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.