Multiscreen® SEQ₃₈₄ Filter Plates
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S384SEQ10 S384SEQ50
MILFS384SEQ10EA 0 EUR
MILFS384SEQ10 MILFS384SEQ50
Multiscreen® SEQ₃₈₄ Filter Plates
Microplaten Filtreerplaten
The MultiScreen384-SEQ filter plate is suited for use with most liquid handling systems. The filter plate operates by vacuum filtration enabling top-loading and recovery of sample. Our plate is designed with the needs of the high throughput laboratory in mind, meets SBS standards and is ideally suited for 'walk away' automation.
  • Automation friendly: Design and protocol saves time, money and improves reproducibility
  • Efficient dye terminator & salt removal: Recovers highly purified sequencing products for long, accurate sequencing reads
  • Eliminates dry-
More Product Information