Millex® Syringe Filters, PTFE Membrane
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SLLG013SL SLLG025SS
MILFSLLG013SLEA 414 EUR
MILFSLLG013SL MILFSLLG025SS
Millex® Syringe Filters, PTFE Membrane
Filters Spuitfilters
Filters for sterilising or clarifying biological solutions. The larger filtration area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
Manufacturing occurs in a controlled environment using an automated process. Provided with a certificate of quality.