Millipore Microscope Slides
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
XX1007615
MILFXX1007615EA 120 EUR
MILFXX1007615
Millipore Microscope Slides
Object- en dekglaasjes Objectglaasjes
A microscope slide is designed for the collection of various samples like the entire specimen, body parts of a specimen, and histological sections for both reference and research purposes.
It has a significant role in history, veterinary, forensic, and medical fields. Microscope slide acts as an accessory for filter holders for sample preparation. Microscope slides have been used for histological studies of necrotic tissue and epithelial debris.