Microplate handling system, StakMax®
Leverancier: Molecular Devices
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
STAKMAX 0200-6016 0200-6017 0200-6187 5005331
MLDVSTAKMAXEA 0 EUR
MLDVSTAKMAX MLDV0200-6016 MLDV0200-6017 MLDV0200-6187 MLDV5005331
Microplate handling system, StakMax®
Microplaatlezers
The StakMax® microplate handling system is a compact benchtop microplate stacker that works with microplate readers and microplate washers to give walk-away automation for up to 50 microplates.
  • Integrates with microplate readers and AquaMax® washers for improved process flow and higher throughput
  • Flexible operation using powerful SoftMax® Pro software
  • Barcode reader can be added to enable barcode identification of plates
  • Minimises plate movement time, while handling the microplates gently, eliminating liquid spillage