Standard safe bucket, DBI-SALA®
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1500140 7100209335
MMMA1500140EA 180 EUR
MMMA1500140 MMMA7100209335
Standard safe bucket, DBI-SALA®
Emmers
DBI-SALA® Safe Bucket is made from vinyl with either a hook and loop or drawstring closure system which helps prevent accidental drops.
  • Heavy-duty vinyl is easy to clean
  • Innovative hook and loop closure system
  • Carabiner and hoisting strap
  • Puncture resistant plating sewn into the base of the bucket
This bucket is designed with six built in connection points and each of them have 4,5 kg load rating.