3M™ PELTOR™ X2B Earmuffs Neckband
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7100123157
MMMA7100123157EA 0 EUR
MMMA7100123157
3M™ PELTOR™ X2B Earmuffs Neckband
Oorkappen Oorkappen met Sound Management
Left and right side attachment conversion kit to earmuffs to Comtac XP and Comtac XPI headsets. Twin headband design provides heat dispersion and internal stainless steel design offers consistent pressure and comfortable fit over an eight hour period.
  • Reduces noise levels up to 31 dB
  • Wide flat headband is electrically insulted making the earmuffs electrically resistant or 'dielectric'
  • Colour-coding makes it easy to identify the level of protection being worn
  • Slim cups offer improved compatibility with other safety equipment
  • Wide soft ear cushions help
More Product Information