Stainless Steel Cleaners
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EDEL600ES EDELS600
MMMAEDEL600ESEA 18 EUR
MMMAEDEL600ES MMMAEDELS600
Stainless Steel Cleaners
Reinigingsmiddelen Detergenten en oppervlaktereinigers
3M™ Stainless steel cleaners are cleaning sprays that create an invisible protective film which slows down the build-up of dirt particles and keeps surfaces cleaner for longer. With a pleasant citrus aroma, they clean, polish, and restore a glossy sheen to stainless steel and other metal surfaces.
  • Polishes and protects all stainless steel surfaces in one step
  • Spray clings to vertical surfaces and leaves a light, non-greasy film that resist fingerprints and marking
  • Does not contain harmful chemicals and is safe to use in food preparation areas
  • Water-based
More Product Information