Scotch-Brite™ 9000 All-Purpose Scouring Pad
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SB9000
MMMASB9000EA 42.3 EUR
MMMASB9000
Scotch-Brite™ 9000 All-Purpose Scouring Pad
Schoonmaakdoekjes Reinigingskits en -pads
The Scotch-Brite™ All-purpose scouring pad 9000 is a thick, hand-sized scouring pad designed for cleaning most types of surfaces without scratching.
  • Versatile, hand-sized pad for dish scrubbing
  • Can be used with Scotch-Brite™ Multi-Purpose Pad Holder 405-R and 3M™ Twist Lok™ Pad Holder 961
Clean most types of surfaces without scratching with Scotch-Brite™ All-purpose scouring pad 9000. Pads are safe for use on porcelain, ceramic, stainless steel, glass, and non-stick pans. It’s specially designed for cleaning hot griddles with mirror or highly polishedMore Product Information