Oppervlaktereiniger, VHB™
Leverancier: 3M
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VHB-1L VHB-25ML
MMMAVHB-1LEA 153 EUR
MMMAVHB-1L MMMAVHB-25ML
Oppervlaktereiniger, VHB™
Reinigingsmiddelen Detergenten en oppervlaktereinigers
VHB™ surface cleaner is a very pure solution of isopropyl alcohol in water. It is colourless, has very little odour, and evaporates very quickly leaving no residue.
  • Degreasing of surfaces
  • Preparation of surfaces for bonding
  • Evaporates quickly leaving no residue
  • Recommended for use with 3M™ VHB™ Tapes
  • Use to create a 'slip plane' effect to allow contact without wear
Apply this cleaner for an odourless, colourless cleaner that evaporates quickly. It leaves no residue when cleaning surfaces. It will remove most light contamination (grease, dirt)More Product Information