Comply Indicator Label
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1322CH
MMME1322CHEA 192 EUR
MMME1322CH
Comply Indicator Label
Sterilisatie-indicatoren
The 3M Comply indicator tapes are an integral part of a coordinated sterilisation control system.
  • Class 1 indicator
  • For sealing packaging
  • Clear colour change
  • Secure closure
  • Readable result
  • can be removed without leaving any residue
The high-quality, stretchable tapes adhere very well to a wide variety of soft packaging.