Time temperature indicators, MonitorMark™
Leverancier: 3M Food Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9860H 9860C 9860D 9861A 9860B
MMMP9860HEA 2180 EUR
MMMP9860H MMMP9860C MMMP9860D MMMP9861A MMMP9860B
Time temperature indicators, MonitorMark™
Temperatuurindicatiestrips
3M™ MonitorMark™ time temperature indicator solves the challenge of monitoring thermal exposure of temperature sensitive products during transportation and storage.
  • Inexpensive solution for monitoring product exposure to a range of temperatures for a specific period of time
  • Self-adhesive backing for easy attachment to secondary packaging
  • Easily-interpreted visual results
  • Results indicate both exposure and relative time over which exposure occurred
  • Removable film activation strip starts the monitoring process on demand for accuracy
These indicators areMore Product Information