Swab-sampler with DE neutralising broth
Leverancier: 3M Food Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
RS96010DE
MMMPRS96010DEEA 146 EUR
MMMPRS96010DE
Swab-sampler with DE neutralising broth
Wattenstaafjes Wattenstaafjes voor monstername
3M™ Swab-Sampler with 10 ml D/E neutralising Broth is a convenient and quick collection device for environmental sampling and testing over small surface areas.
  • D/E neutralising Broth vial is ready to use for pathogen testing of environmental samples
  • Swab reaches into corners and hard-to-reach areas for increased accuracy
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant
  • Bottles have ample headspace for sample addition and vortexing
  • Self-contained swab and vial for increased productivity and efficiency