Blood glucose lancets, VITREX® Soft
Leverancier: Vitrex Medical A/S
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
351213 352213 354213 357213
MODU351213EA 15 EUR
MODU351213 MODU352213 MODU354213 MODU357213
Blood glucose lancets, VITREX® Soft
Bloedlancetten
Needle material is stainless steel grade 304/1 Cr18Ni9 and moulded body/cap material is LDPE (Low Density Polyethylene).
  • Good blood flow - Triple ground needle point
  • Sterilised by gamma irradiation at minimum 25kGy
  • Convenient boxes with dispensing feature
VITREX® Soft is designed for use with most popular types of finger lancing device.