Smart® FFP Masks with Comfortable Nose Seal
Leverancier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
238501
MOLD238501EA 83.2 EUR
MOLD238501
Smart® FFP Masks with Comfortable Nose Seal
Ademhalingsmaskers Wegwerpmaskers Stofmaskers
Smart FFP masks are designed with a soft foam nose cushion for a comfortable fit. A soft, skin-friendly inner fleece layer and a stretchy woven strap provide additional comfort for the wearer.
  • Reliable protection against particles
  • Soft nose cushion for enhanced wearing comfort
  • ActivForm® 3D design for a comfortable and secure fit
The flexible ActivForm® 3D design adapts automatically to the shape of the wearers face and their facial movements. As a result, our Smart masks are easy to wear and sit securely and comfortably on the skin at all times,More Product Information