Pura-Fit® Classic Foam Earplugs
Leverancier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
776001
MOLD776001EA 119 EUR
MOLD776001
Pura-Fit® Classic Foam Earplugs
Oordoppen Oordoppen voor eenmalig gebruik
The classic Pura-Fit® earplugs offer the optimal combination of stability and softness. They are easy to roll and, upon insertion, expand to fill the ear canal at exactly the right moment. There, they maintain a secure, sealed fit, without exerting pressure on the ear.
  • Noise reduction at high (H), medium (M) and low (L) frequencies
  • 36 dB (SNR) noise reduction
  • Soft foam for a high degree of wearing comfort
  • Also available in a dispenser and with neck cord
Shelf life from date of production: 4 years.
Order Now
Specificaties
Environmentally Preferable