Culture media, ready to use, in tubes

Leverancier: Dr. Möller & Schmelz
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

4040 5200 4045 4080 5220 5150 4090 4020 4021 4025 5130 4030 4047 4050 4051 4055 4060 4061 4062 4096 4100 4125 4130 4135 4150 5140 5160 5170 5241 5251 5281
391-3880EA 67 EUR
391-3880 MOLL5200 MOLL4045 MOLL4080 MOLL5220 MOLL5150 MOLL4090 MOLL4020 MOLL4021 MOLL4025 MOLL5130 MOLL4030 MOLL4047 MOLL4050 MOLL4051 MOLL4055 MOLL4060 MOLL4061 MOLL4062 MOLL4096 MOLL4100 MOLL4125 MOLL4130 MOLL4135 MOLL4150 MOLL5140 MOLL5160 MOLL5170 MOLL5241 MOLL5251 MOLL5281
Culture media, ready to use, in tubes
Media voor microbiologie Geprepareerde media voor microbiologie

These sterile ready-to-use media are filled in autoclavable and break-proof glass tubes (20 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C they can be poured as agar plates in different sizes and thicknesses to meet individual demands.


  • Delivered 50 ml in polycarbonate bottles or 20 ml in glass tubes

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR