Drum taps, PE
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2801 2802
MSGA2801EA 28.8 EUR
MSGA2801 MSGA2802
Drum taps, PE
Kranen en gootstenen Kranen
These polyethylene taps are used for corrosive and aggressive chemicals.
  • Fast outflow
  • Tri-sure thread