Stackable plastic trays
Leverancier: MULTIROIR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
26500PENPR 26501PENPR 26502PENPR 26503PENPR 26505PENPR 26506PENPR
MULC26500PENPREA 99 EUR
MULC26500PENPR MULC26501PENPR MULC26502PENPR MULC26503PENPR MULC26505PENPR MULC26506PENPR
Stackable plastic trays
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Range of six stackable plastic trays for storage.
  • Made from polypropylene
  • Stackable
  • High impact resistance
  • Six colors available
Trays are suitable for use in temperatures ranging from −20 to 60 °C.