Nile red, Technical Grade, Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

M-Clarity™ quality level = MQ200

N3013-100MG N3013-1G
N3013-100MGEA 187 EUR
N3013-100MG N3013-1G
Nile red, Technical Grade, Sigma-Aldrich®
Nile red
Formule: C₂₀H₁₈N₂O₂
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00011639
CAS nummer: 7385-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc6d204d74ada3ce080a08e3e9633ea0096ba9b1&vid=20929756552703bea970d64f897edb14b6d9c050&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Faint Green to Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon 70.0 - 77.3 %
Nitrogen 7.5 - 8.8 %
Solubility (Colour) Faint Pink to Very Dark Red
Solubility (Turbidity)
1 mg/mL, MeOH
Clear to Turbid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR