Sluitingen voor flessen, InVitro™, Biotainer®, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2560-0489 2560-0490
NALG2560-0489EA 216 EUR
NALG2560-0489 NALG2560-0490
Sluitingen voor flessen, InVitro™, Biotainer®, Nalgene®
Sluitingen Flesdoppen
Designed to aseptically handle solutions in biopharmaceutical, biotechnology and laboratory liquid transfer applications, these two- and three-ported closures are designed for Thermo Scientific™ Nalgene™ Biotainer™ bottles and carboys. With Animal Derived Component-Free (ADCF) components, including seals that are sonically-welded to the cap, and leak-proof closures, security of high value contents are guaranteed during fluid transfer.
  • With ADCF components
  • Including seals that are sonically welded to the cap
  • Leak-proof closures
  • Security of high value
More Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement