Flessen, brede hals, HDPE, rechthoekig, met schroefdop, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
312007-0008 312007-0016 312007-0004 312007-0032
NALG312007-0008EA 688 EUR
NALG312007-0008 NALG312007-0016 NALG312007-0004 NALG312007-0032
Flessen, brede hals, HDPE, rechthoekig, met schroefdop, Nalgene®
Flessen Kunststof flessen
HDPE, translucent, with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof
  • Non sterile
Ideal for collecting, storing and shipping samples or storing liquids and dry substances. Space saving rectangular base.
Certificaten: Manufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.