Slangen, PFA, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8051-0125 8051-0187 8051-0250 8051-0375 8051-0500
NALG8051-0125EA 126 EUR
NALG8051-0125 NALG8051-0187 NALG8051-0250 NALG8051-0375 NALG8051-0500
Slangen, PFA, Nalgene®
Slangen Transfer Tubing
Made of PFA, translucent.
  • Semi-rigid with very high chemical resistance
  • Wide temperature range: –270 to 260 °C
  • Durometer hardness: Shore D, 55
  • Very low water absorption
For low-pressure use with mechanical, compression-type fittings. Can be sterilized by autoclaving, gas, dry heat, and disinfectants. Do not sterilize by radiation.
Certificaten: Resin complies with USDA regulations.