Spore ampoules
Leverancier: CROSSTEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SA1-15-06 SA1-50-06 SA1-15-04 SA1-50-04 SA1-15-05 SA1-50-05 SA1-NC-10
489-0113EA 288 EUR
489-0113 489-0114 NAMSSA1-15-04 NAMSSA1-50-04 NAMSSA1-15-05 NAMSSA1-50-05 NAMSSA1-NC-10
Spore ampoules
Sterilisatie-indicatoren
These spore ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles. Spore ampoules are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic
Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. No activation is required; simply expose Spore Ampoules to sterilization process and incubate.
  • Reduced incubation time of 48 hours
  • Clear visible results as growth is evident by turbidity and/or a color shift of the media from purple
More Product Information