Whirl-Pak® Cellulose Sponge Bags
Leverancier: NASCO FORT ATKINSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B01245 B01423 B01422
NASCB01245EA 305 EUR
NASCB01245 NASCB01423 NASCB01422
Whirl-Pak® Cellulose Sponge Bags
Zakken Monsterzakken
Cellulose sponges for your environmental monitoring and surface sampling needs.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labelling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against leaks
  • Tape extends beyond wire to eliminate sharp ends and provide a
More Product Information