Graduated pipettes for enzyme test, Class AS
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0262-05-011 0262-07-011 0262-07-022 0262-07-051 0262-07-101
NEUB0262-05-011EA 26.8 EUR
NEUB0262-05-011 NEUB0262-07-011 NEUB0262-07-022 NEUB0262-07-051 NEUB0262-07-101
Graduated pipettes for enzyme test, Class AS
Pipetten Gegradueerde pipetten
Graduated pipettes with long pulled out tip, ring divisions and partial sequence above O-point.
Certificaten: DIN 12699; Class AS, conformity certified.