Measuring cylinders, tall form, class B, borosilicate glass
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0284-95-0250 0284-95-0103 0284-95-0253
NEUB0284-95-0250EA 93.1 EUR
NEUB0284-95-0250 NEUB0284-95-0103 NEUB0284-95-0253
Measuring cylinders, tall form, class B, borosilicate glass
Cilinders Maatcilinders
Borosilicate glass, tall form, class B.
  • Round glass base with spout
  • Blue graduations
Certificaten: DIN 53194