Laboratory bottles, pressure resistant, glass
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0854-22-2000 0854-11-0100 0854-11-2000 0854-11-300045 0854-11-050004 0854-11-500045 0854-121000045 0854-16-5045
NEUB0854-22-2000EA 379 EUR
NEUB0854-22-2000 NEUB0854-11-0100 NEUB0854-11-2000 NEUB0854-11-300045 NEUB0854-11-050004 NEUB0854-11-500045 NEUB0854-121000045 NEUB0854-16-5045
Laboratory bottles, pressure resistant, glass
Flessen Glazen flessen
These bottles are made from pressure resistant borosilicate glass 3.3.
  • Without cap
  • Without spout
Pressure resistant from –1 to 1,5 bar
Certificaten: ISO 4796.