Canada Balsam Glass Jar
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7298-50-50
NEUB7298-50-50EA 198 EUR
NEUB7298-50-50
Canada Balsam Glass Jar
Potjes Potjes voor algemeen gebruik
Canada balsam glass, according to Goldschmidt, is used for microscopy and comes with a glass stirring rod.