Wall-mount mat roll dispensers, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MAT292
NEWPMAT292EA 136 EUR
NEWPMAT292
Wall-mount mat roll dispensers, PIG®
Dispensers voor doekjes
These mat roll dispensers feature a braking mechanism for more controlled dispensing.
  • Easy to refill and replace
  • Slot and cork brake prevents mat from unfurling as it's dispensed
  • Durable hot-rolled steel frame and galvanised core rod built to last in industrial settings
Order Now
Specificaties
Environmentally Preferable