PIG® Overpack Dolly
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PAK414
NEWPPAK414EA 251 EUR
NEWPPAK414
PIG® Overpack Dolly
Transportwagens
The wide-base design helps ensure stability during transport of overpacks or rapid response with a PIG overpack spill kit.
  • Heavy-duty rolled steel construction holds up to 363 kg
  • Lockable casters keep overpack stationary, even on uneven surfaces