Spill containment trays, SW series
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PAKE274SW PAKE271SW-YW PAKE273SW-YW PAKE270SWD
NEWPPAKE274SWEA 51.6 EUR
NEWPPAKE274SW NEWPPAKE271SW-YW NEWPPAKE273SW-YW NEWPPAKE270SWD
Spill containment trays, SW series
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Lightweight and durable PE spill trays are ideal for storing several small to medium sized containers of liquids.
  • Chemical and rust resistant
  • Stackable for easy transportation and storage
  • Standard trays with high and durable sidewalls; containers sit deep into the sump, minimising any potential splashes
  • Spill trays with grate feature an anti-slip, removable grate that keeps containers away from collected liquids
Opgelet: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammableMore Product Information