PIG® Original absorbent socks
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
404 204 PIG205 PIG209 PIG218 PIG219 PIG220
NEWP404EA 128 EUR
NEWP404 NEWP204 NEWPPIG205 NEWPPIG209 NEWPPIG218 NEWPPIG219 NEWPPIG220
PIG® Original absorbent socks
Absorptiematerialen en bescherming tegen morsen
Formable socks hug corners and surround machine bases to absorb machine leaks, contain puddles and help keep workers safe.
  • Tough and economical; good choice for everyday leak protection
  • Contained corncob filler pulls liquids from the floor for maximum absorbency whilst making cleanup quick and easy
  • Polypropylene skin resists tearing; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water
  • Corncob filler is light-weight and economical to handle general, everyday tasks
Can be incinerated after use toMore Product Information