Wipers for general applications, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WIP235 WIP230 WIP231 WIP310 WIP311
NEWPWIP235EA 72 EUR
NEWPWIP235 NEWPWIP230 NEWPWIP231 NEWPWIP310 NEWPWIP311
Wipers for general applications, PIG®
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Doekjes voor algemeen gebruik
These cellulose wipes with adhesive latex binder are ideal for all kinds of jobs - from cleaning parts to cleaning hands.
  • Constructed of soft, absorbent cellulose fibers with a high-quality binder to increase strength for wet or dry wiping
  • Made thick and absorbent to quickly soak up oils, degreasers and other liquids
  • Low-lint material won't leave residue or create a mess
  • Excellent for a variety of wiping applications from quick cleanups to degreasing
  • Available 1/4 folded and poly wrapped, as economical rolls or in convenient pop-up boxes
Order Now
Specificaties
Environmentally Preferable