BioClean™ Sterile Bags
Leverancier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S-BBPN-1 S-BBPN-3 S-BBPN-14 S-BBPN-4 S-BBPN-15 S-BBPN-6
NITRS-BBPN-1EA 0 EUR
NITRS-BBPN-1 NITRS-BBPN-3 NITRS-BBPN-14 NITRS-BBPN-4 NITRS-BBPN-15 NITRS-BBPN-6
BioClean™ Sterile Bags
Zakken Opbergzakken
Manufactured in a cleanroom environment from ultra-clean low-density polythene. Because the material is free of additives, there is no contamination from particulates, surface leaching chemicals or out-gassing.
  • Gamma sterilized
  • Double bagged
  • Range of sizes available
  • Method: Gamma irradiation
  • Minimum Dose: 25 kGy
  • Sterility Assurance Level: 10⁻⁶
For general-purpose use, ideal as waste bags, transportation bags or storage bags. BioClean™ Sterile Bags are suitable for use within the pharmaceutical, medical, semiconductor, aerospace, and many otherMore Product Information